Please login to your online account to display your discounted pricing

anti-HSP27, Clone: G3.1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone G3.1
Host Species Mouse
Isotype κ
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Antigen HSP27
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232972

 
novus biologicals canada
NBP232972
Unlabeled 0.5mL Each for $321.00

NBP2329721M

 
novus biologicals canada
NBP2329721ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone G3.1
Host Species Mouse
Isotype κ
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Antigen HSP27
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Chimpanzee.
Gene Accession No. P04792
Gene ID (Entrez) 3315
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL