Please login to your online account to display your discounted pricing

anti-HSP27, Clone: G3.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329721ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329721M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Cross Reactivity Sheep
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 3315
Conjugate Unlabeled
Format Purified
Clone G3.1
Isotype κ
Regulatory Status RUO
Test Specificity Chimpanzee.
Gene Accession No. P04792
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Immunogen Partially purified hsp27 (earlier called 24K) protein from breast cancer MCF-7 cells.
Quantity 1mL
Antigen HSP27