Please login to your online account to display your discounted pricing

anti-HPV16 L1, Clone: CamVir 1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002732SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002732SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P03101
Formulation PBS
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG2a
Test Specificity Cross reacts with HPV37. Reacts very strongly with biopsy specimins containing HPV16 or HPV33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV6 or HPV11. Human Papillomavirus (HPV).
Host Species Mouse
Immunogen A beta galactosidase-L1 fusion protein purified by PAGE.
Cross Reactivity Virus
Quantity 0.025mL
Clone CamVir 1
Antigen HPV16 L1
Format Purified
Regulatory Status RUO