Please login to your online account to display your discounted pricing

anti-HPV16 L1, Clone: CamVir 1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1002732

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1002732

  • / Each
 

Description & Specifications

Specifications

Antigen HPV16 L1
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone CamVir 1
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Virus
Format Purified
Formulation PBS
Gene Accession No. P03101
Host Species Murine
Immunogen A beta galactosidase-L1 fusion protein purified by PAGE.
Isotype IgG2a
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Cross reacts with HPV37. Reacts very strongly with biopsy specimins containing HPV16 or HPV33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV6 or HPV11. Human Papillomavirus (HPV).
Target Species Virus