Please login to your online account to display your discounted pricing

anti-HPV16 L1, Clone: CamVir 1, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone CamVir 1
Host Species Mouse
Isotype IgG2a
Immunogen A beta galactosidase-L1 fusion protein purified by PAGE.
Antigen HPV16 L1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1002732SS

 
novus biologicals canada
NB1002732SS
Unlabeled 0.025mL Each for $139.00

NB1002732

 
novus biologicals canada
NB1002732
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone CamVir 1
Host Species Mouse
Isotype IgG2a
Immunogen A beta galactosidase-L1 fusion protein purified by PAGE.
Antigen HPV16 L1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Cross reacts with HPV37. Reacts very strongly with biopsy specimins containing HPV16 or HPV33; very weak reactions were occasionally observed with biopsy specimens or smears containing HPV6 or HPV11. Human Papillomavirus (HPV).
Gene Accession No. P03101
Formulation PBS
Concentration 1.0mg/mL