Please login to your online account to display your discounted pricing

anti-HPi2, Clone: HIC1-2B4.2B, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone HIC1-2B4.2B
Host Species Murine
Isotype IgG1
Immunogen Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells.
Antigen HPi2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP118946SS

 
novus biologicals canada
NBP118946SS
Unlabeled 0.025mL Each for $139.00

NBP118946

 
novus biologicals canada
NBP118946
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone HIC1-2B4.2B
Host Species Murine
Isotype IgG1
Immunogen Human pancreatic enriched islet cells containing low levels of exocrine and ductal cells.
Antigen HPi2
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Test Specificity Specific for multiple endocrine cell types.
Concentration 1mg/mL