Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Clone: 425D11.01/ clone 3, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX1306005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX1306005

  • / Each
 

Description & Specifications

Specifications

Isotype IgG1
Regulatory Status RUO
Cross Reactivity Virus
Antigen HIV-1 gp41
Formulation Antibody in 100μg in 200μL/50μg in 100μL Tris-NaCl pH 8
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Immunogen 92BR025
Quantity 0.05mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone 425D11.01/ clone 3
Conjugate Unlabeled
Format Ion Exchange Chromatography