Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Clone: 425D11.01/ clone 3, Novus Biologicals™

$309.05 - $586.39

Specifications

Clone 425D11.01/ clone 3
Host Species Mouse
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX1306

 
novus biologicals canada
DDX1306
Unlabeled 0.1mg Each for $586.39

DDX1306005

 
novus biologicals canada
DDX1306005MG
Unlabeled 0.05mg Each for $309.05
Description & Specifications

Specifications

Clone 425D11.01/ clone 3
Host Species Mouse
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Formulation Antibody in 100μg in 200μL/50μg in 100μL Tris-NaCl pH 8