Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Clone: 417D3.08/ clone 2, Novus Biologicals™

$309.05 - $586.39

Specifications

Clone 417D3.08/ clone 2
Host Species Murine
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX1305

 
novus biologicals canada
DDX1305
Unlabeled 0.1mg Each for $586.39

DDX1305005

 
novus biologicals canada
DDX1305005MG
Unlabeled 0.05mg Each for $309.05
Description & Specifications

Specifications

Clone 417D3.08/ clone 2
Host Species Murine
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Formulation Antibody in 100μg in 200μL/50μg in 100μL Tris-NaCl pH 8