Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Clone: 417D3.08/ clone 2, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX1305005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX1305005

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Primary or Secondary Primary
Conjugate Unlabeled
Formulation Antibody in 100μg in 200μL/50μg in 100μL Tris-NaCl pH 8
Immunogen 92BR025
Quantity 0.05mg
Regulatory Status RUO
Antigen HIV-1 gp41
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Virus
Format Ion Exchange Chromatography
Isotype IgG1
Clone 417D3.08/ clone 2