Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Clone: 402D9.14/clone 1, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX1304005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX1304005

  • / Each
 

Description & Specifications

Specifications

Antigen HIV-1 gp41
Applications ELISA
Applications Flow Cytometry
Clone 402D9.14/clone 1
Conjugate Unlabeled
Cross Reactivity Virus
Format Ion Exchange Chromatography
Formulation Antibody at 0.5mg/mL in Tris-NaCl pH 8
Host Species Murine
Immunogen 92BR025
Isotype IgG1
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Target Species Virus