Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Clone: 402D9.14/clone 1, Novus Biologicals™

$329.00 - $586.39

Specifications

Clone 402D9.14/clone 1
Host Species Mouse
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX1304005

 
novus biologicals canada
DDX1304005MG
Unlabeled 0.05mg Each for $329.00

DDX1304

 
novus biologicals canada
DDX1304
Unlabeled 0.1mg Each for $586.39
Description & Specifications

Specifications

Clone 402D9.14/clone 1
Host Species Mouse
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Formulation Antibody at 0.5mg/mL in Tris-NaCl pH 8