Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Biotin, Clone: 425D11.01/ clone 3, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX1306B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX1306B005

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Isotype IgG1
Primary or Secondary Primary
Cross Reactivity Virus
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Conjugate Biotin
Formulation Antibody in 100μg in 200μL/50μg in 100μL PBS 50% glycerol
Format Ion Exchange Chromatography
Applications ELISA
Applications Flow Cytometry
Antigen HIV-1 gp41
Clone 425D11.01/ clone 3
Immunogen 92BR025
Quantity 0.05mg