Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Biotin, Clone: 425D11.01/ clone 3, Novus Biologicals™

$406.00 - $659.00

Specifications

Clone 425D11.01/ clone 3
Host Species Mouse
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX1306B

 
novus biologicals canada
DDX1306B
Biotin 0.1mg Each for $659.00

DDX1306B005

 
novus biologicals canada
DDX1306B005MG
Biotin 0.05mg Each for $406.00
Description & Specifications

Specifications

Clone 425D11.01/ clone 3
Host Species Mouse
Isotype IgG1
Immunogen 92BR025
Antigen HIV-1 gp41
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Formulation Antibody in 100μg in 200μL/50μg in 100μL PBS 50% glycerol