Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Biotin, Clone: 417D3.08/ clone 2, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX1305B

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX1305B

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Conjugate Biotin
Monoclonal or Polyclonal Monoclonal
Formulation Antibody in 100μg in 200μL/50μg in 100μL PBS 50% glycerol
Format Ion Exchange Chromatography
Cross Reactivity Virus
Applications ELISA
Applications Flow Cytometry
Clone 417D3.08/ clone 2
Antigen HIV-1 gp41
Primary or Secondary Primary
Immunogen 92BR025
Quantity 0.1mg
Isotype IgG1
Regulatory Status RUO