Please login to your online account to display your discounted pricing

anti-HIV-1 gp41, Biotin, Clone: 417D3.08/ clone 2, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX1305B005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX1305B005

  • / Each
 

Description & Specifications

Specifications

Antigen HIV-1 gp41
Immunogen 92BR025
Primary or Secondary Primary
Host Species Mouse
Clone 417D3.08/ clone 2
Format Ion Exchange Chromatography
Isotype IgG1
Applications ELISA
Applications Flow Cytometry
Formulation Antibody in 100μg in 200μL/50μg in 100μL PBS 50% glycerol
Conjugate Biotin
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Virus
Regulatory Status RUO
Quantity 0.05mg