Please login to your online account to display your discounted pricing

anti-Histone H4 (Monomethyl Lys20), Clone: 5E10-D8, Novus Biologicals™

$109.00 - $362.00

Specifications

Clone 5E10-D8
Host Species Murine
Isotype κ
Immunogen Branched peptide found between amino acids 10-100 of Histone H4. [UniProt No. P62805]
Antigen Histone H4 (Monomethyl Lys20)
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP130091SS

 
novus biologicals canada
NBP130091SS
Unlabeled 0.025mL Each for $109.00

NBP130091

 
novus biologicals canada
NBP130091
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Clone 5E10-D8
Host Species Murine
Isotype κ
Immunogen Branched peptide found between amino acids 10-100 of Histone H4. [UniProt No. P62805]
Antigen Histone H4 (Monomethyl Lys20)
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity All mammals including human, mouse and rat, Drosophila, C. elegans, yeast.
Gene Accession No. P62805
Gene ID (Entrez) 121504
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL