Please login to your online account to display your discounted pricing

anti-Histone H3 (p Ser28), Clone: 117C826, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056595SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056595T

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide containing phospho serine at position 28 CARKS*APATGGVKK, of human Histone H3 was used as immunogen. The immunogen sequence (CARKS*APATGGVKK) is conserved across mammalian and invertebrates, as well as plants.
Formulation PBS
Conjugate Unlabeled
Gene Accession No. Q71DI3
Primary or Secondary Primary
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 126961
Test Specificity In anti-Fas mAb treated Jurkat, an 18 kDa band is observed. Reacts in Human. Predicted to react with most Mammals and Invertebrates.
Cross Reactivity Human
Format Purified
Host Species Mouse
Clone 117C826
Isotype IgG1
Regulatory Status RUO
Applications Western Blotting
Quantity 0.025mg
Antigen Histone H3 (p Ser28)