Please login to your online account to display your discounted pricing

anti-Histone H3 (p Ser28), Clone: 117C826, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056595

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056595

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide containing phospho serine at position 28 CARKS*APATGGVKK, of human Histone H3 was used as immunogen. The immunogen sequence (CARKS*APATGGVKK) is conserved across mammalian and invertebrates,as well as plants.
Formulation PBS
Primary or Secondary Primary
Gene ID (Entrez) 126961
Test Specificity In anti-Fas mAb treated Jurkat, an 18 kDa band is observed. Reacts in Human. Predicted to react with most Mammals and Invertebrates.
Conjugate Unlabeled
Gene Accession No. Q71DI3
Applications Western Blotting
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Isotype IgG1
Clone 117C826
Cross Reactivity Human
Format Purified
Antigen Histone H3 (p Ser28)
Host Species Mouse
Regulatory Status RUO