Please login to your online account to display your discounted pricing

anti-HIP1 Related, Clone: 1E1, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 1.00E+01
Host Species Murine
Isotype κ
Immunogen Human HIP1 Related-TALIN Homology Region (57 kDa). [UniProt No. O75146]
Antigen HIP1 Related
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300206SS

 
novus biologicals canada
NB300206SS
Unlabeled 0.025mL Each for $109.00

NB300206

 
novus biologicals canada
NB300206
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 1.00E+01
Host Species Murine
Isotype κ
Immunogen Human HIP1 Related-TALIN Homology Region (57 kDa). [UniProt No. O75146]
Antigen HIP1 Related
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. O75146
Gene ID (Entrez) 9026