Please login to your online account to display your discounted pricing

anti-Hepatocyte Specific Antigen, Clone: HSA98, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HSA98
Host Species Mouse
Isotype κ
Immunogen HEP-3B human hepatocellular carcinoma cells
Antigen Hepatocyte Specific Antigen
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328601M

 
novus biologicals canada
NBP2328601ML
Unlabeled 1mL Each for $429.00

NBP232860

 
novus biologicals canada
NBP232860
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone HSA98
Host Species Mouse
Isotype κ
Immunogen HEP-3B human hepatocellular carcinoma cells
Antigen Hepatocyte Specific Antigen
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL