Please login to your online account to display your discounted pricing

anti-Hepatocyte Specific Antigen, Clone: HSA133, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328611ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328611M

  • / Each
 

Description & Specifications

Specifications

Antigen Hepatocyte Specific Antigen
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Clone HSA133
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen SK-H1A9-2 human hepatocellular carcinoma cells
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal