Please login to your online account to display your discounted pricing

anti-Hepatocyte Specific Antigen, Clone: HSA133, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232861

 View all options for this product


For Research Use Only

Catalog No. NBP232861


Description & Specifications


Antigen Hepatocyte Specific Antigen
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Clone HSA133
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen SK-H1A9-2 human hepatocellular carcinoma cells
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal