Please login to your online account to display your discounted pricing

anti-Hepatocyte Specific Antigen, Clone: HSA133, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HSA133
Host Species Mouse
Isotype κ
Immunogen SK-H1A9-2 human hepatocellular carcinoma cells
Antigen Hepatocyte Specific Antigen
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232861

 
novus biologicals canada
NBP232861
Unlabeled 0.5mL Each for $321.00

NBP2328611M

 
novus biologicals canada
NBP2328611ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone HSA133
Host Species Mouse
Isotype κ
Immunogen SK-H1A9-2 human hepatocellular carcinoma cells
Antigen Hepatocyte Specific Antigen
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL