Please login to your online account to display your discounted pricing

anti-hCG alpha, Clone: HCGa/53, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232970

 View all options for this product


For Research Use Only

Catalog No. NBP232970


Description & Specifications


Antigen hCG alpha
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone HCGa/53
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01215
Host Species Mouse
Immunogen Recombinant hCG alpha protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1081