Please login to your online account to display your discounted pricing

anti-hCG alpha, Clone: HCGa/53, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HCGa/53
Host Species Mouse
Isotype κ
Immunogen Recombinant hCG alpha protein
Antigen hCG alpha
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329701M

 
novus biologicals canada
NBP2329701ML
Unlabeled 1mL Each for $429.00

NBP232970

 
novus biologicals canada
NBP232970
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone HCGa/53
Host Species Mouse
Isotype κ
Immunogen Recombinant hCG alpha protein
Antigen hCG alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. P01215
Gene ID (Entrez) 1081
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL