Please login to your online account to display your discounted pricing

anti-hCG alpha, Clone: HCGa/53, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329701M

  • / Each
 

Description & Specifications

Specifications

Antigen hCG alpha
Applications ELISA
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone HCGa/53
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01215
Host Species Mouse
Immunogen Recombinant hCG alpha protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1081