Please login to your online account to display your discounted pricing

anti-Golgi Complex, Clone: 371-4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328521ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328521M

  • / Each
 

Description & Specifications

Specifications

Antigen Golgi Complex
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen SU-DHL-1 large cell lymphoma cells.
Isotype κ
Host Species Mouse
Primary or Secondary Primary
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Quantity 1mL
Clone 371-4
Format Purified