Please login to your online account to display your discounted pricing

anti-Golgi Complex, Clone: 371-4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232852

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232852

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Quantity 0.5mL
Clone 371-4
Conjugate Unlabeled
Antigen Golgi Complex
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen SU-DHL-1 large cell lymphoma cells.
Isotype κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Regulatory Status RUO