Please login to your online account to display your discounted pricing

anti-Golgi Complex, Clone: 371-4, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 371-4
Host Species Mouse
Isotype κ
Immunogen SU-DHL-1 large cell lymphoma cells.
Antigen Golgi Complex
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232852

 
novus biologicals canada
NBP232852
Unlabeled 0.5mL Each for $321.00

NBP2328521M

 
novus biologicals canada
NBP2328521ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 371-4
Host Species Mouse
Isotype κ
Immunogen SU-DHL-1 large cell lymphoma cells.
Antigen Golgi Complex
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL