Please login to your online account to display your discounted pricing

anti-Mouse GM-CSF, Polyclonal, R&D Systems

$467.98 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q14AD9
Gene ID (Entrez) 12981
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AB415NA

 
r & d systems
RDSAB415NA
Unlabeled 1mg Each for $467.98

AF415NA

 
r & d systems
RDSAF415NA
Unlabeled 100μg Each for $467.98
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q14AD9
Gene ID (Entrez) 12981

Host Species: Goat
Species Reactivity:Mouse
Immunogen:E. coli-derived recombinant mouse GM-CSF, Ala18-Lys141, Accession No. Q14AD9

Applications:

Neutralization, Western Blotting