Please login to your online account to display your discounted pricing

anti-Glucose 6 Phosphate Dehydrogenase, Clone: 2H7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222125

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222125

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Immunogen Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P11413]
Isotype IgG1
Format Precipitation
Host Species Mouse
Regulatory Status RUO
Cross Reactivity Human
Antigen Glucose 6 Phosphate Dehydrogenase
Clone 2H7
Concentration 1.0mg/mL
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Formulation PBS
Gene Accession No. P11413
Gene ID (Entrez) 2539
Primary or Secondary Primary