Please login to your online account to display your discounted pricing

anti-Glucose 6 Phosphate Dehydrogenase, Clone: 2H7, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 2H7
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P11413]
Antigen Glucose 6 Phosphate Dehydrogenase
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222125SS

 
novus biologicals canada
NBP222125SS
Unlabeled 0.025mL Each for $139.00

NBP222125

 
novus biologicals canada
NBP222125
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Clone 2H7
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P11413]
Antigen Glucose 6 Phosphate Dehydrogenase
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P11413
Gene ID (Entrez) 2539
Formulation PBS
Concentration 1.0mg/mL