Please login to your online account to display your discounted pricing

anti-Glucose 6 Phosphate Dehydrogenase, Clone: 2H7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222125SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222125SS

  • / Each
 

Description & Specifications

Specifications

Immunogen Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P11413]
Quantity 0.025mL
Clone 2H7
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Regulatory Status RUO
Cross Reactivity Human
Antigen Glucose 6 Phosphate Dehydrogenase
Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Format Precipitation
Formulation PBS
Host Species Mouse
Gene Accession No. P11413
Primary or Secondary Primary
Gene ID (Entrez) 2539
Conjugate Unlabeled