Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: NB615, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300285SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300285SS

  • / Each
 

Description & Specifications

Specifications

Antigen GAPDH
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Clone NB615
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Supernatant
Gene Accession No. P04406
Host Species Murine
Immunogen Full length native GAPDH from human erythrocytes. [UniProt No. P04406]
Isotype IgM
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2597
Target Species Bovine
Target Species Canine
Target Species Chicken
Target Species Human
Target Species Murine
Target Species Porcine
Target Species Rabbit
Target Species Rat