Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: NB615, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300285

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300285

  • / Each
 

Description & Specifications

Specifications

Format Supernatant
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Quantity 0.1mL
Clone NB615
Gene ID (Entrez) 2597
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Host Species Mouse
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P04406
Isotype IgM
Immunogen Full length native GAPDH from human erythrocytes. [UniProt No. P04406]
Antigen GAPDH