Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: NB615, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone NB615
Host Species Murine
Isotype IgM
Immunogen Full length native GAPDH from human erythrocytes. [UniProt No. P04406]
Antigen GAPDH
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300285

 
novus biologicals canada
NB300285
Unlabeled 0.1mL Each for $309.05

NB300285SS

 
novus biologicals canada
NB300285SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone NB615
Host Species Murine
Isotype IgM
Immunogen Full length native GAPDH from human erythrocytes. [UniProt No. P04406]
Antigen GAPDH
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Gene Accession No. P04406
Gene ID (Entrez) 2597