Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: 6C5, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600502

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600502

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Applications Immunoassay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Isotype IgG1
Cross Reactivity Amphibian
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Fish
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human or rabbit GAPDH.
Primary or Secondary Primary
Gene ID (Entrez) 2597
Formulation PBS. pH 7.4
Monoclonal or Polyclonal Monoclonal
Test Specificity Bovine (Negative), Goat (Negative).
Gene Accession No. P04406
Conjugate Unlabeled
Format Purified
Antigen GAPDH
Clone 6C5
Regulatory Status RUO