Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: 6C5, Novus Biologicals™

$109.00 - $252.00

Specifications

Clone 6C5
Host Species Murine
Isotype IgG1
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human or rabbit GAPDH.
Antigen GAPDH
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600502

 
novus biologicals canada
NB600502
Unlabeled 0.1mg Each for $252.00

NB600502SS

 
novus biologicals canada
NB600502SS
Unlabeled 0.02mg Each for $109.00
Description & Specifications

Specifications

Clone 6C5
Host Species Murine
Isotype IgG1
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human or rabbit GAPDH.
Antigen GAPDH
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoassay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Test Specificity Bovine (Negative), Goat (Negative).
Gene Accession No. P04406
Gene ID (Entrez) 2597
Formulation PBS. pH 7.4