Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: 6C5, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600502SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600502SS

  • / Each
 

Description & Specifications

Specifications

Applications Immunoassay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human or rabbit GAPDH.
Cross Reactivity Amphibian
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Fish
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rat
Regulatory Status RUO
Gene ID (Entrez) 2597
Isotype IgG1
Host Species Mouse
Quantity 0.02mg
Formulation PBS. pH 7.4
Gene Accession No. P04406
Primary or Secondary Primary
Test Specificity Bovine (Negative), Goat (Negative).
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Antigen GAPDH
Format Purified
Clone 6C5