Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: 2D4A7, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300328

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300328

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Isotype κ
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Quantity 0.1mL
Clone 2D4A7
Antigen GAPDH
Immunogen Ni-NTA purified recombinant human GAPDH expressed in &italic_begin;E. coli&italic_end; strain BL21 (DE3). [UniProt No. P04406]
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2597
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P04406
Primary or Secondary Primary
Host Species Mouse
Test Specificity Human, Mouse, Rabbit and Rat. Since GAPDH is one of the most conserved proteins known and it is therefore likely that the antibody will react with other species also. Porcine reactivity reported in scientific literature (PMID: 25063310).
Concentration 1.0mg/mL
Regulatory Status RUO