Please login to your online account to display your discounted pricing

anti-GAPDH, Clone: 2D4A7, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 2D4A7
Host Species Murine
Isotype κ
Immunogen Ni-NTA purified recombinant human GAPDH expressed in &italic_begin;E. coli&italic_end; strain BL21 (DE3). [UniProt No. P04406]
Antigen GAPDH
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB300328

 
novus biologicals canada
NB300328
Unlabeled 0.1mL Each for $329.00

NB300328SS

 
novus biologicals canada
NB300328SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 2D4A7
Host Species Murine
Isotype κ
Immunogen Ni-NTA purified recombinant human GAPDH expressed in &italic_begin;E. coli&italic_end; strain BL21 (DE3). [UniProt No. P04406]
Antigen GAPDH
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Human, Mouse, Rabbit and Rat. Since GAPDH is one of the most conserved proteins known and it is therefore likely that the antibody will react with other species also. Porcine reactivity reported in scientific literature (PMID: 25063310).
Gene Accession No. P04406
Gene ID (Entrez) 2597
Formulation Tris-glycine, 150mM NaCl
Concentration 1.0mg/mL