Please login to your online account to display your discounted pricing

anti-GAP-43, Clone: 1E3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP192713SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP192713SS

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Clone 1.00E+03
Format Purified
Regulatory Status RUO
Antigen GAP-43
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Quantity 0.025mL
Immunogen C-terminal peptide of rat and mouse GAP43 (KEDPEADQEHA) to which an N-terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
Host Species Mouse
Primary or Secondary Primary
Gene ID (Entrez) 2596
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Formulation PBS