Please login to your online account to display your discounted pricing

anti-GAP-43, Clone: 1E3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP192713

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP192713

  • / Each
 

Description & Specifications

Specifications

Antigen GAP-43
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Clone 1.00E+03
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS
Host Species Murine
Immunogen C-terminal peptide of rat and mouse GAP43 (KEDPEADQEHA) to which an N-terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
Isotype κ
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2596
Target Species Human
Target Species Murine
Target Species Rat