Please login to your online account to display your discounted pricing

anti-GAP-43, Clone: 1E3, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone 1.00E+03
Host Species Murine
Isotype κ
Immunogen C-terminal peptide of rat and mouse GAP43 (KEDPEADQEHA) to which an N-terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
Antigen GAP-43
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP192713

 
novus biologicals canada
NBP192713
Unlabeled 0.1mL Each for $309.05

NBP192713SS

 
novus biologicals canada
NBP192713SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 1.00E+03
Host Species Murine
Isotype κ
Immunogen C-terminal peptide of rat and mouse GAP43 (KEDPEADQEHA) to which an N-terminal Cysteine residue was added to allow chemical coupling to Keyhole Limpet Hemocyanin carrier protein.
Antigen GAP-43
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Gene ID (Entrez) 2596
Formulation PBS
Concentration 1mg/mL