Please login to your online account to display your discounted pricing

anti-FoxP3, Clone: 3G3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056582SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056582T

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Formulation PBS containing 0.2% gelatin
Conjugate Unlabeled
Gene ID (Entrez) 50943
Concentration 0.5mg/mL
Gene Accession No. Q9BZS1
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Applications Flow Cytometry
Applications Western Blotting
Antigen FoxP3
Immunogen Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known
Quantity 0.025mg
Clone 3G3
Host Species Mouse
Isotype κ
Format Purified