Please login to your online account to display your discounted pricing

anti-FoxP3, Clone: 3G3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056582

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056582

  • / Each
 

Description & Specifications

Specifications

Antigen FoxP3
Applications Flow Cytometry
Applications Western Blot
Clone 3G3
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.2% gelatin
Gene Accession No. Q9BZS1
Host Species Murine
Immunogen Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 50943
Target Species Human
Target Species Murine