Please login to your online account to display your discounted pricing

anti-FoxO1/FKHR, Clone: 83N7F8, Novus Biologicals™

$109.00 - $307.00

Specifications

Clone 83N7F8
Host Species Murine
Isotype κ
Immunogen FOXO1A
Antigen FoxO1/FKHR
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP231376

 
novus biologicals canada
NBP231376
Unlabeled 0.1mg Each for $307.00

NBP231376SS

 
novus biologicals canada
NBP231376SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 83N7F8
Host Species Murine
Isotype κ
Immunogen FOXO1A
Antigen FoxO1/FKHR
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity Human. Based upon immunogen sequence simlarity, this antibody is predicted to react with Mouse (92%), Rat (92%), Bovine/Cow (91%), Squirrel (91%) and Porcine/Pig (90%).
Gene Accession No. Q12778
Gene ID (Entrez) 2308
Formulation PBS
Concentration 1.0mg/mL