Please login to your online account to display your discounted pricing

anti-FosB/G0S3, Clone: 83B1138, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 83B1138
Host Species Murine
Isotype IgG1
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB.
Antigen FosB/G0S3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056530

 
novus biologicals canada
NB10056530
Unlabeled 0.1mg Each for $329.00

NB10056530T

 
novus biologicals canada
NB10056530SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 83B1138
Host Species Murine
Isotype IgG1
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB.
Antigen FosB/G0S3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In HeLa, a 46 kDa band is observed.
Gene Accession No. P53539
Gene ID (Entrez) 2354
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL