Please login to your online account to display your discounted pricing

anti-FosB/G0S3, Clone: 83B1138, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056530SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056530T

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Gene Accession No. P53539
Conjugate Unlabeled
Applications Western Blotting
Isotype IgG1
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2354
Test Specificity In HeLa, a 46 kDa band is observed.
Concentration 0.5mg/mL
Cross Reactivity Human
Format Purified
Host Species Mouse
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 2-15 of human FosB.
Antigen FosB/G0S3
Regulatory Status RUO
Clone 83B1138
Quantity 0.025mg